missing translation for 'onlineSavingsMsg'
Learn More

2-Demethyl Colchicine, TRC

Product Code. 30012385
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012385 10 mg 10mg
30008350 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012385 Supplier Toronto Research Chemicals Supplier No. TRCD23088010MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102491-80-5
Molecular Formula C21 H23 N O6
Molecular Weight (g/mol) 385.41
IUPAC Name N-[(7S)-2-hydroxy-1,3,10-trimethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
SMILES COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(O)c(OC)c23)NC(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H23NO6/c1-11(23)22-15-7-5-12-9-18(27-3)20(25)21(28-4)19(12)13-6-8-17(26-2)16(24)10-14(13)15/h6,8-10,15,25H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1
Formula Weight 385.1525
Chemical Name or Material 2-Demethyl Colchicine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.