missing translation for 'onlineSavingsMsg'
Learn More

2’-Deoxy-beta-L-adenosine (May contain up to 20% inorganics), TRC

Product Code. 30354138
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354138 50 mg 50mg
30366997 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30354138 Supplier Toronto Research Chemicals Supplier No. TRCD23162850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Deoxy-beta-L-adenosine
CAS 14365-45-8
Molecular Formula C10 H13 N5 O3
Quantity 50 mg
Synonym 9-(2-Deoxy-β-L-erythro-pentofuranosyl)-9H-purin-6-amine Hydrochloride
InChI Formula InChI=1S/C10H13N5O3/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(17)6(2-16)18-7/h3-7,16-17H,1-2H2,(H2,11,12,13)/t5-,6+,7+/m1/s1
SMILES Nc1ncnc2c1ncn2[C@@H]3C[C@@H](O)[C@H](CO)O3
IUPAC Name (2S,3R,5S)-5-(6-aminopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol) 251.24
Formula Weight 251.1018
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.