missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-galactose, TRC

Product Code. 30366790
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30366790 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30366790

Brand: Toronto Research Chemicals TRCD2370001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Deoxy-D-galactose
CAS 1949-89-9
Molecular Formula C6 H12 O5
Quantity 1 g
Synonym D-lyxo-Hexose, 2-deoxy-, 2-Deoxy-D-lyxo-hexose, Galactose, 2-deoxy-, D- (6CI), 2-Deoxy-D-galactose, 2-Deoxy-D-talose, 2-Deoxygalactose, Galactose, 2-deoxy-, (3R,4R,5R)-3,4,5,6-Tetrahydroxyhexanal
InChI Formula InChI=1S/C6H12O5/c7-2-4-6(10)3(8)1-5(9)11-4/h3-10H,1-2H2/t3-,4-,5?,6-/m1/s1
SMILES OC[C@H]1OC(O)C[C@@H](O)[C@H]1O
IUPAC Name (4R,5R,6R)-6-(hydroxymethyl)oxane-2,4,5-triol
Molecular Weight (g/mol) 164.16
Formula Weight 164.0685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.