missing translation for 'onlineSavingsMsg'
Learn More

2-Deoxy-D-ribose-13C5, TRC

Product Code. 30179366
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.50mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179366 25 mg 25mg
30179056 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179366 Supplier Toronto Research Chemicals Supplier No. TRCD25200125MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 266998-43-0
Molecular Formula 13C5 H10 O4
Molecular Weight (g/mol) 139.09
Synonym D-erythro-Pentose-1,2,3,4,5-13C5, 2-deoxy-, 2-Deoxy-D-ribose-13C5, [1,2,3,4,5-13C5]-2-Deoxy-D-ribose, D-erythro-Pentose-13C5, 2-deoxy- (9CI), 2-Deoxy-D-erythro-pentose-1,2,3,4,5-13C5
IUPAC Name (3S,4R)-3,4,5-trihydroxy(1,2,3,4,5-^{13}C_{5})pentanal
SMILES O[13CH2][13C@@H](O)[13C@@H](O)[13CH2][13CH]=O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H10O4/c6-2-1-4(8)5(9)3-7/h2,4-5,7-9H,1,3H2/t4-,5+/m0/s1/i1+1,2+1,3+1,4+1,5+1
Formula Weight 139.0747
Chemical Name or Material 2-Deoxy-D-ribose-13C5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.