missing translation for 'onlineSavingsMsg'
Läs mer

Thermo Scientific Chemicals 2'-Deoxyadenosine-5'-triphosphate disodium salt, 97%

Produktkod. 15410457
Change view
Klicka för att se tillgängliga alternativ
Quantity:
500 mg
Förpackningsstorlek:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
15410457 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15410457 Leverantör Thermo Scientific Chemicals Leverantörsnummer J65542.ME
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 74299-50-6
Color White
Molecular Formula C10H16N5O12P3
MDL Number MFCD00080335,MFCD00167019,MFCD11164415
Quantity 500 mg
Synonym datp,disodium salt,trihydrate
InChI Key SUYVUBYJARFZHO-UHFFFAOYNA-N
SMILES NC1=C2N=CN(C3CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O3)C2=NC=N1
IUPAC Name [[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] phosphono hydrogen phosphate;sodium;trihydrate
Molecular Weight (g/mol) 491.18
PubChem CID 129893645
Formula Weight 535.14
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2'-Deoxyadenosine-5'-triphosphate disodium salt
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.