missing translation for 'onlineSavingsMsg'
Learn More

2-(Di-p-tolylphosphino)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10785452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10785452 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10785452 Supplier Thermo Scientific Acros Supplier No. 435155000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1202865-03-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 109.0°C to 110.5°C
Color Yellow
Elemental Analysis 76.05 to 82.40% C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C21H19OP
MDL Number MFCD15143661
Quantity 500 mg
Synonym 2-di-p-tolylphosphino benzaldehyde, 2-bis 4-methylphenyl phosphanyl benzaldehyde, acmc-20aon9, 2-bis 4-methylphenyl phosphino benzaldehyde, 2-di-p-tolylphosphino benzaldehyde,min
InChI Key IDDRQJVTYBKJLP-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=CC=CC=C3C=O
IUPAC Name 2-bis(4-methylphenyl)phosphanylbenzaldehyde
Molecular Weight (g/mol) 318.35
PubChem CID 71310894
Formula Weight 318.35
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-(Di-p-Tolylphosphino)benzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Di-p-tolylphosphino)benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.