Learn More
2-(Di-tert-butylphosphino)ethylamine, 10 wt.% solution in THF, Thermo Scientific™
Product Code. 11915951 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 mL
Packaging:
Glass bottle
Unit Size:
2.5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11915951 2.5 mL Glass bottle 2.5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11915951 Supplier Thermo Scientific Acros Supplier No. 435030025
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
CAS 1053658-84-6, 109-99-9
Color Colorless to Tan
Density 0.8800g/mL
Flash Point −21°C
Packaging Glass bottle
Molecular Formula C10H24NP
Linear Formula ((CH3)3C)2PCH2CH2NH2
Quantity 2.5 mL
Specific Gravity 0.88
Synonym 2-di-t-butylphosphino ethylamine, 2-di-tert-butylphosphino ethylamine, 2-ditertbutylphosphino ethylamine, 2-aminoethyldi-tert-butylphosphine, 2-di-tert-butylphosphino ethanamine, 2-di-tert-butylphosphanyl ethan-1-amine, 2-aminoethyl di-tert-butyl phosphane, 2-di-tert-butylphosphino ethylamine, 10 wt. % in thf
InChI Key DZDGRVHIUYIMDS-UHFFFAOYSA-N
SMILES CC(C)(C)P(CCN)C(C)(C)C
IUPAC Name 2-ditert-butylphosphanylethanamine
Molecular Weight (g/mol) 189.28
PubChem CID 57553001
Formula Weight 189.28
Percent Purity 9 to 11%
Physical Form Solution
Chemical Name or Material 2-(Di-tert-butylphosphino)ethylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-(Di-tert-butylphosphino)ethylamine, 10wt.% solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H250-Catches fire spontaneously if exposed to air.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.