missing translation for 'onlineSavingsMsg'
Learn More

2-(Diethylamino)ethyl 4-(2-Hydroxybenzamido)benzoate, TRC

Product Code. 30388128
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388128 1 mg
30376508 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388128 Supplier Toronto Research Chemicals Supplier No. TRCD4435601MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1007652-84-7
Molecular Formula C20H24N2O4
Molecular Weight (g/mol) 356.42
Synonym 2-(Diethylamino)ethyl Ester 4-[(2-Hydroxybenzoyl)amino]benzoic Acid, Procaine Impurity, N-Desmethyl O-Desoctyl Otilonium
SMILES CCN(CC)CCOC(=O)c1ccc(NC(=O)c2ccccc2O)cc1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H24N2O4/c1-3-22(4-2)13-14-26-20(25)15-9-11-16(12-10-15)21-19(24)17-7-5-6-8-18(17)23/h5-12,23H,3-4,13-14H2,1-2H3,(H,21,24)
Formula Weight 356.1736
Chemical Name or Material 2-(Diethylamino)ethyl 4-(2-Hydroxybenzamido)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.