Learn More
2-Docosahexaenoin-1-oleoyl 3-Phosphocholine, TRC
Product Code. 30369903
Change view
Click to view available options
Quantity:
5 mg
50 mg
Product Code. Quantity
30369903 50 mg
30370333 5 mg
2 options
Product Code. 30369903
Brand: Toronto Research Chemicals TRCD49448050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 99296-82-9
Molecular Formula C48H82NO8P
Molecular Weight (g/mol) 832.14
Synonym [R-(all-Z)]-4-Hydroxy-N,N,N-trimethyl-9-oxo-7-[[[(9Z)-1-oxo-9- octadecen-1-yl]oxy]methyl]-3,5,8-trioxa-4-phosphatriaconta-12,15,18,21,24,27-hexaen-1-aminium Inner Salt 4-Oxide
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OC[C@H](COP(=O)([O-])OCC[N+](C)(C)C)OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C48H82NO8P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-39-41-48(51)57-46(45-56-58(52,53)55-43-42-49(3,4)5)44-54-47(50)40-38-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h8,10,14,16,20-22,24-26,29,31,35,37,46H,6-7,9,11-13,15,17-19,23,27-28,30,32-34,36,38-45H2,1-5H3/b10-8-,16-14-,22-20-,25-24-,26-21-,31-29-,37-35-/t46-/m1/s1
Chemical Name or Material 2-Docosahexaenoin-1-oleoyl 3-Phosphocholine