missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-6,9-diaminoacridine Lactate, TRC

Product Code. 30383935
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30383935 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383935 Supplier Toronto Research Chemicals Supplier No. TRCE2615051G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethacridine Lactate
CAS 1837-57-6
Molecular Formula C15 H15 N3 O . C3 H6 O3
Quantity 1 g
Synonym Propanoic acid, 2-hydroxy-, compd. with 7-ethoxy-3,9-acridinediamine (1:1), 6,9-Diamino-2-ethoxyacridine lactate, Lactic acid compd. with 6,9-diamino-2-ethoxyacridine (1:1), 7-Ethoxy-3,9-acridinediamine 2-hydroxypropanoate (1:1), 6,9-Diamino-2-ethoxyacrid
InChI Formula InChI=1S/C15H15N3O.C3H6O3/c1-2-19-10-4-6-13-12(8-10)15(17)11-5-3-9(16)7-14(11)18-13;1-2(4)3(5)6/h3-8H,2,16H2,1H3,(H2,17,18);2,4H,1H3,(H,5,6)
SMILES CCOc1ccc2nc3cc(N)ccc3c(N)c2c1.CC(O)C(=O)O
IUPAC Name 7-ethoxyacridine-3,9-diamine;2-hydroxypropanoic acid
Molecular Weight (g/mol) 343.377
Formula Weight 343.153
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.