missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine, 94%, Thermo Scientific™

Product Code. 11357525
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11357525

Brand: Thermo Scientific Alfa Aesar L05649.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14529-53-4
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00023464
InChI Key LISKAOIANGDBTB-UHFFFAOYSA-N
Synonym pyridine, 2-ethoxy, 2-ethoxy pyridine, 2-ethoxy-pyridine, zlchem 304, pubchem1126, acmc-209cun, ethyl 2-pyridinyl ether, ksc497c7d
PubChem CID 84499
IUPAC Name 2-ethoxypyridine
SMILES CCOC1=CC=CC=N1

Specifications

Density 1.009
Boiling Point 155°C to 156°C
Flash Point 52°C (126°F)
Refractive Index 1.496
Quantity 50 g
UN Number UN1993
Beilstein 111115
Formula Weight 123.16
Percent Purity 94%
Chemical Name or Material 2-Ethoxypyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.