missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-indanone, TRC

Product Code. 30382428
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382428 100 mg 100mg
30379170 50 mg -
30371972 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382428 Supplier Toronto Research Chemicals Supplier No. TRCE937208100MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22351-56-0
Molecular Formula C11H12O
Molecular Weight (g/mol) 160.21
SMILES CCC1Cc2ccccc2C1=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C11H12O/c1-2-8-7-9-5-3-4-6-10(9)11(8)12/h3-6,8H,2,7H2,1H3
Formula Weight 160.0888
Chemical Name or Material 2-Ethyl-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.