missing translation for 'onlineSavingsMsg'
Learn More

(2-Ethylhexyl)magnesium bromide, 1M solution in diethyl ether, AcroSeal™

Product Code. 11905971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11905971 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11905971 Supplier Thermo Scientific Acros Supplier No. 435951000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 90224-21-8 | C8H17BrMg | 217.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90224-21-8, 60-29-7
CAS Min % 23.94, 72.29
CAS Max % 27.71, 76.07
Color Colorless to Gray
Density 0.8630g/mL
Flash Point −25°C
Packaging AcroSeal™ Glass Bottle
Molecular Formula C8H17BrMg
MDL Number MFCD02260220
Quantity 100 mL
Specific Gravity 0.863
Synonym 2-ethylhexylmagnesium bromide, 2-ethylhexyl magnesium bromide, 2-ethylhexylmagensium bromide, 2-ethylhexylmagnesiumbromide, 2-ethylhexyl-magnesiumbromide, 2-ethylhexylmagnesium-bromide, 2-ethylhexyl-magnesium bromide, ndcjrlbsqqdnoo-uhfffaoysa-m, grignard reagent
Solubility Information Solubility in water: reacts.
InChI Key NDCJRLBSQQDNOO-UHFFFAOYNA-M
SMILES CCCCC(CC)C[Mg]Br
IUPAC Name bromo(2-ethylhexyl)magnesium
Molecular Weight (g/mol) 217.43
PubChem CID 14915511
Formula Weight 217.43
Physical Form Solution
Chemical Name or Material (2-Ethylhexyl)magnesium bromide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • (2-Ethylhexyl)magnesium bromide, 1M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.