Learn More
2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Product Code. 15437456
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437456 250 mg 250mg
15427456 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15437456 Supplier Thermo Scientific Alfa Aesar Supplier No. H62451.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1256360-51-6
Molecular Formula C17H25BFNO3
Molecular Weight (g/mol) 321.199
InChI Key VSRFPDFCDOCPEQ-UHFFFAOYSA-N
Synonym 4-2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl morpholine, 2-fluoro-3-morpholinomethyl phenylboronic acid, pinacol ester, 4-2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine, 4-2-fluoro-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl morpholine
PubChem CID 53217280
IUPAC Name 4-[[2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]morpholine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)CN3CCOCC3)F
Specifications
Quantity 250 mg
Formula Weight 321.19
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester