missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzoic acid, 99%, ACROS Organics™

Product Code. 10297270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10297270 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10297270 Supplier Thermo Scientific Acros Supplier No. 308180010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 115029-24-8
CAS Min % 98.5
CAS Max % 100.0
Melting Point 165°C to 169°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C8H4F4O2
Linear Formula FC6H3(CF3)COH
MDL Number MFCD00010322
Quantity 1 g
Synonym 2-fluoro-4-trifluoromethyl benzoic acid, 2-fluoro-4-trifluoromethyl benzoicacid, alpha,alpha,alpha,2-tetrafluoro-p-toluic acid, benzoic acid, 2-fluoro-4-trifluoromethyl, acmc-20aidb, pubchem2079, ksc509c3f, rarechem al bo 0632, 4-carboxy-3-fluorobenzotrifluoride
InChI Key OCIYTBZXTFPSPI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)F)C(=O)O
IUPAC Name 2-fluoro-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol) 208.112
PubChem CID 518003
Formula Weight 208.11
Percent Purity ≥98.5%
Physical Form Crystals
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzoic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.