missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%, BLD Pharm™

Product Code. 30783105
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
250mg
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783105 100 g 100g
30780908 1 g 1g
30768364 5 g 5g
30775322 10 g 10g
30773329 250 mg 250mg
30773308 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30783105 missing translation for 'mfr' BLD Pharm missing translation for 'supplierNo' BD10791100g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 146070-34-0
Molecular Formula C8H3F4N
MDL Number MFCD00061279
Quantity 100 g
SMILES N#CC1=CC=C(C(F)(F)F)C=C1F
Molecular Weight (g/mol) 189.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-(trifluoromethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.