missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-bromopyridine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 2-Fluoro-5-bromopyridine |
| CAS | 766-11-0 |
| Molecular Formula | C5H3BrFN |
| Quantity | 10 g |
| Synonym | 3-Bromo-6-fluoropyridine, 5-Bromo-2-fluoropyridine |
| InChI Formula | InChI=1S/C5H3BrFN/c6-4-1-2-5(7)8-3-4/h1-3H |
| SMILES | Fc1ccc(Br)cn1 |
| IUPAC Name | 5-bromo-2-fluoropyridine |
| Molecular Weight (g/mol) | 175.99 |
| Formula Weight | 174.9433 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction