Learn More
2-Fluorobenzoic Acid, TRC
Product Code. 30405251
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405251 10 g
30404307 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30405251 Supplier Toronto Research Chemicals Supplier No. TRCF58838010G
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 445-29-4
Molecular Formula C7H5FO2
Molecular Weight (g/mol) 140.11
Synonym o-Fluorobenzoic Acid, NSC 10319
SMILES OC(=O)c1ccccc1F
Specifications
Quantity 10 g
InChI Formula InChI=1S/C7H5FO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)
Formula Weight 140.0274
Chemical Name or Material 2-Fluorobenzoic Acid