missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenethyl alcohol, 99%, Thermo Scientific™

Product Code. 10482261 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10482261 5 g Glass bottle 5g
10237710 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10482261 Supplier Thermo Scientific Acros Supplier No. 310970050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 50919-06-7
Molecular Formula C8H9FO
Molecular Weight (g/mol) 140.16
MDL Number MFCD00002887
InChI Key HNIGZVZDWCTFPR-UHFFFAOYSA-N
Synonym 2-fluorophenethyl alcohol, 2-2-fluorophenyl ethanol, 2-2-fluorophenyl ethan-1-ol, benzeneethanol, 2-fluoro, 2-fluorophenethylalcohol, 2-2-fluoro-phenyl-ethanol, 2-fluorobenzeneethanol, acmc-1axdv, o-fluorophenethyl alcohol, 2-fluorophenylethyl alcohol
PubChem CID 2733296
IUPAC Name 2-(2-fluorophenyl)ethanol
SMILES OCCC1=CC=CC=C1F

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0400g/mL
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4CH2CH2OH
Refractive Index 1.5080 to 1.51
Quantity 5 g
Specific Gravity 1.04
Formula Weight 140.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Fluorophenethyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.