missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropropionic Acid, TRC

Product Code. 30398718
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30398718 5 g 5g
30396695 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398718 Supplier Toronto Research Chemicals Supplier No. TRCF5986105G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6087-13-4
Molecular Formula C3 H5 F O2
Molecular Weight (g/mol) 92.07
Synonym 2-Fluoropropanoic Acid, (±)-2-Fluoropropionic Acid, NSC 84352, α-Fluoropropionic Acid
IUPAC Name 2-fluoropropanoic acid
SMILES CC(F)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H5FO2/c1-2(4)3(5)6/h2H,1H3,(H,5,6)
Formula Weight 92.0274
Chemical Name or Material 2-Fluoropropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.