missing translation for 'onlineSavingsMsg'
Learn More

2'-Fluoropropiophenone, 99%

Product Code. 11348795
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11348795

Brand: Thermo Scientific Alfa Aesar L12423.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 446-22-0 | C9H9FO | 152.17 g/mol

2'-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoropropiophenone is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in chloroform, methanol. Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-22-0
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD00009893
InChI Key NSNSIFGTEGKZFK-UHFFFAOYSA-N
Synonym 2'-fluoropropiophenone, 1-2-fluorophenyl propan-1-one, 1-2-fluorophenyl-1-propanone, ethyl 2-fluorophenyl ketone, 1-propanone, 1-2-fluorophenyl, 1-2-fluorphenyl propan-1-on, 2;-fluoropropiophenone, pubchem19917, acmc-1cpyw, o-fluoropropiophenone
PubChem CID 579399
IUPAC Name 1-(2-fluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=CC=C1F

Specifications

Density 1.102
Boiling Point 89°C to 91°C (12 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.5045
Quantity 2 g
Beilstein 3235723
Solubility Information Soluble in chloroform,methanol. Not miscible in water.
Formula Weight 152.17
Percent Purity 99%
Chemical Name or Material 2'-Fluoropropiophenone
compliance-icons
Product Identifier
  • 2'-Fluoropropiophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2'-Fluoropropiophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.