missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinopyrimidine, 97%

Product Code. 15791739 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15791739 5 g 5g
15781739 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15791739 Supplier Thermo Scientific Acros Supplier No. 453050050
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7504-94-1 | C4H6N4 | 110.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7504-94-1
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD01249337
InChI Key QDGHXQFTWKRQTG-UHFFFAOYSA-N
Synonym 2-hydrazinylpyrimidine, 2-hydrazinopyrimidine, 2-hydrazino-pyrimidine, pyrimidin-2-yl-hydrazine, pyrimidine-2-ylhydrazine, 2 1h-pyrimidinone, hydrazone, 2 1h-pyrimidinone, hydrazone 9ci, hydrazinopyrimidine, aminoiminopyrimidine, 2-hydrazinopyrimdine
PubChem CID 346558
IUPAC Name pyrimidin-2-ylhydrazine
SMILES NNC1=NC=CC=N1

Specifications

Melting Point 110.0°C to 112.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 2-Hydrazinopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.