missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-(carboxymethylamino)-hydrocinnamic Acid, Dipotassium Salt, TRC

Product Code. 30394948
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30394948 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394948 Supplier Toronto Research Chemicals Supplier No. TRCH886125500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-3-(carboxymethylamino)-hydrocinnamic Acid, Dipotassium Salt
CAS 100750-38-7
Molecular Formula C11H11K2NO5
Quantity 500 mg
InChI Formula InChI=1S/C11H13NO5.2K/c13-9(14)5-4-7-2-1-3-8(11(7)17)12-6-10(15)16;;/h1-3,12,17H,4-6H2,(H,13,14)(H,15,16);;/q;2*+1/p-2
SMILES [K+].[K+].Oc1c(CCC(=O)[O-])cccc1NCC(=O)[O-]
IUPAC Name dipotassium;3-[3-(carboxylatomethylamino)-2-hydroxyphenyl]propanoate
Molecular Weight (g/mol) 315.41
Formula Weight 314.9911
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.