Learn More
Alfa Aesar™ 2-Hydroxy-4,6-dimethylpyrimidine, 97%
Product Code. 11470284
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11470284 25 g 25g
11480284 100 g 100g
2 options
Product Code. 11470284
Brand: Alfa Aesar™ A13676.14
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Chemical Identifiers
CAS 108-79-2
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD00006072
InChI Key WHEQVHAIRSPYDK-UHFFFAOYSA-N
Synonym 2-hydroxy-4,6-dimethylpyrimidine, 4,6-dimethylpyrimidin-2-ol, 4,6-dimethyl-2-hydroxypyrimidine, 4,6-dimethylpyrimidin-2 1h-one, 2 1h-pyrimidinone, 4,6-dimethyl, unii-hg0cbn4v9f, 4,6-dimethyl-2-pyrimidinol, 4,6-dimethyl-pyrimidin-2-ol, hg0cbn4v9f, 4,6-dimethyl-2-pyrimidone
PubChem CID 9508
IUPAC Name 4,6-dimethyl-1H-pyrimidin-2-one
SMILES CC1=CC(C)=NC(=O)N1
Specifications
Melting Point 202°C to 204°C
Quantity 25 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4,6-dimethylpyrimidine