missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylbenzaldehyde, TRC

Product Code. 30394754
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
missing translation for 'unitSize':
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30394754 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30394754 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCH5050201G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxy-4-methylbenzaldehyde
CAS 698-27-1
Molecular Formula C8H8O2
Quantity 1 g
InChI Formula InChI=1S/C8H8O2/c1-6-2-3-7(5-9)8(10)4-6/h2-5,10H,1H3
SMILES Cc1ccc(C=O)c(O)c1
IUPAC Name 2-hydroxy-4-methylbenzaldehyde
Molecular Weight (g/mol) 136.15
Formula Weight 136.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.