Learn More
2'-Hydroxy-5'-methylacetophenone, 98%
Description
2'-Hydroxy-5'-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2′-Hydroxy-5′-methylacetophenone is used to produce 2-ethyl-4-methyl-phenol. It is used as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 45°C to 48°C |
| Density | 1.079 |
| Boiling Point | 112°C to 114°C (12 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 5 g |
| Beilstein | 1364840 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 150.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 2'-Hydroxy-5'-methylacetophenone |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.