missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-methylbenzophenone, ACROS Organics™

Product Code. 10686934 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10686934 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10686934 Supplier Thermo Scientific Acros Supplier No. 121430010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 1470-57-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 83°C to 85°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C14H12O2
Linear Formula HOC6H3(CH3)COC6H5
MDL Number MFCD00002379
Quantity 1 g
Beilstein 08, 177
Synonym 2-hydroxy-5-methylbenzophenone, 2-benzoyl-4-methylphenol, 2-hydroxy-5-methylphenyl phenyl methanone, unii-85w615bmp7, methanone, 2-hydroxy-5-methylphenyl phenyl, 2-hydroxy-5-methyl-benzophenone, 2-hydroxy-5-methylphenyl-phenylmethanone, acmc-209cy0, dsstox_cid_27857, dsstox_rid_82610
InChI Key OQERFUGURPLBQH-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1)O)C(=O)C2=CC=CC=C2
IUPAC Name (2-hydroxy-5-methylphenyl)-phenylmethanone
Molecular Weight (g/mol) 212.248
PubChem CID 73851
Formula Weight 212.25
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-5-methylbenzophenone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.