missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitropyridine, 98+%

Product Code. 11461638
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461638 25 g 25g
11471638 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461638 Supplier Thermo Scientific Alfa Aesar Supplier No. A13921.14
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5418-51-9 | C5H4N2O3 | 140.098 g/mol

A comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A comparison of catalysts promotes imidazolide couplings including the identification of 2-hydroxy-5-nitropyridine as a new, safe, and effective catalyst.

Solubility
Soluble in hot water and alkali liquor, Insoluble in most of organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5418-51-9
Molecular Formula C5H4N2O3
Molecular Weight (g/mol) 140.098
MDL Number MFCD00006276
InChI Key XKWSQIMYNVLGBO-UHFFFAOYSA-N
Synonym 2-hydroxy-5-nitropyridine, 5-nitropyridin-2-ol, 5-nitro-2-pyridinol, 2 1h-pyridinone, 5-nitro, 5-nitropyridin-2 1h-one, 5-nitro-2-hydroxypyridine, 5-nitro-2-pyridone, 5-nitro-1,2-dihydropyridin-2-one, 5-nitro-2-pyridol, pubchem1205
PubChem CID 79453
IUPAC Name 5-nitro-1H-pyridin-2-one
SMILES C1=CC(=O)NC=C1[N+](=O)[O-]

Specifications

Melting Point 188°C to 191°C
Quantity 25 g
Beilstein 120352
Solubility Information Soluble in hot water and alkali liquor,Insoluble in most of organic solvents.
Formula Weight 140.1
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-5-nitropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.