missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzimidazole, 98%

Product Code. 11493887
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493887 250 g 250g
11473887 10 g 10g
11483887 50 g 50g
11309609 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11493887 Supplier Thermo Scientific Alfa Aesar Supplier No. A18324.30
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 615-16-7 | C7H6N2O | 134.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-16-7
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD00127894
InChI Key SILNNFMWIMZVEQ-UHFFFAOYSA-N
Synonym 2-hydroxybenzimidazole, 2-benzimidazolol, 2-benzimidazolinone, o-phenyleneurea, 2-benzimidazolone, 2-oxobenzimidazole, 2 3h-benzimidazolone, 1,3-dihydro-2h-benzimidazol-2-one, 2h-benzimidazol-2-one, 1,3-dihydro, benzamidazole-2 3h-one
PubChem CID 11985
SMILES O=C1NC2=CC=CC=C2N1

Specifications

Melting Point 314°C to 316°C
Quantity 250 g
Beilstein 119866
IUPAC Name 2,3-dihydro-1H-1,3-benzodiazol-2-one
Formula Weight 134.14
Percent Purity 98%
Chemical Name or Material 2-Hydroxybenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.