missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzimidazole, TRC

Product Code. 30179856
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179856 10 g 10g
30180128 100 g 100g
30180302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179856 Supplier Toronto Research Chemicals Supplier No. TRCH82900010G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-16-7
Molecular Weight (g/mol) 134.14
Synonym 2H-Benzimidazol-2-one, 1,3-dihydro-, 2-Benzimidazolinone (6CI,7CI,8CI), 1,3-Dihydro-2H-benzimidazol-2-one, 1,3-Dihydro-2H-benzimidazole-2-one, 1,3-Dihydrobenzimidazol-2-one, 1H-Benzimidazol-2(3H)-one, 1H-Benzo[d]imidazol-2(3H)-one, 2(3H)-Benzimidazolone, 2(3H)-Oxobenzimidazole, 2,3-Dihydro-2-oxo-1H-benzimidazole, 2-Benzimidazolol, 2-Benzimidazolone, 2-Hydroxybenzimidazole, N,N'-(1,2-Phenyleneurea), NSC 10383, NSC 178108, Urea, N,N'-(1,2-phenylene)-, o-Phenyleneurea, Lansoprazole Imp. D (EP)
IUPAC Name 1,3-dihydrobenzimidazol-2-one
SMILES O=C1Nc2ccccc2N1

Specifications

Molecular Formula C7 H6 N2 O
Quantity 10 g
InChI Formula InChI=1S/C7H6N2O/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)
Formula Weight 134.048
Chemical Name or Material 1,3-Dihydro-2H-benzimidazol-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.