missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxybenzimidazole, TRC

Product Code. 30180302
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180302 1 g 1g
30179856 10 g 10g
30180128 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180302 Supplier Toronto Research Chemicals Supplier No. TRCH8290001G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-16-7
Molecular Weight (g/mol) 134.14
SMILES O=C1Nc2ccccc2N1

Specifications

Molecular Formula C7H6N2O
Quantity 1 g
Chemical Name or Material 2-Hydroxybenzimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.