missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxychrysene, TRC

Product Code. 30137301
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137301 10 mg 10mg
30135688 2.5 mg 2.5mg
30005572 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137301 Supplier Toronto Research Chemicals Supplier No. TRCH90534010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65945-06-4
Molecular Formula C18H12O
Molecular Weight (g/mol) 244.29
Synonym 2-Chrysenol
SMILES OC1=CC(C=CC2=C3C=CC4=CC=CC=C24)=C3C=C1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H12O/c19-14-7-10-16-13(11-14)6-9-17-15-4-2-1-3-12(15)5-8-18(16)17/h1-11,19H
Formula Weight 244.0888
Chemical Name or Material 2-Hydroxychrysene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.