missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxydecanedioic Acid, TRC

Product Code. 30393220
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393220 50 mg
30398363 10 mg
30391592 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393220 Supplier Toronto Research Chemicals Supplier No. TRCH93945050MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103963-71-9
Molecular Formula C10H18O5
Molecular Weight (g/mol) 218.25
Synonym alpha-Hydroxysebacic Acid
IUPAC Name 2-hydroxydecanedioic acid
SMILES OC(CCCCCCCC(=O)O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H18O5/c11-8(10(14)15)6-4-2-1-3-5-7-9(12)13/h8,11H,1-7H2,(H,12,13)(H,14,15)
Formula Weight 218.1154
Chemical Name or Material 2-Hydroxydecanedioic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.