missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl acrylate, 97%, stab. with 200-300ppm 4-methoxyphenol

Product Code. 11446576
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446576 1000 g 1000g
11456576 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446576 Supplier Thermo Scientific Alfa Aesar Supplier No. B24815.0B
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 818-61-1 | C5H8O3 | 116.116 g/mol

As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, peroxides, metals, copper and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-61-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO

Specifications

Melting Point <-60°C
Density 1.106
Boiling Point 192°C
Flash Point 101°C (214°F)
Odor Pungent
Linear Formula CH2=CHCO2CH2CH2OH
Refractive Index 1.45
Quantity 1000 g
UN Number UN2927
Sensitivity Light sensitive
Solubility Information Miscible with water.
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material 2-Hydroxyethyl acrylate, stabilized with 200 to 300ppm 4-methoxyphenol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Hydroxyethyl acrylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Hydroxyethyl acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.