missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzaldehyde, 98%

Product Code. 11318416
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318416 1 g 1g
11328416 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318416 Supplier Thermo Scientific Alfa Aesar Supplier No. L17446.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26260-02-6 | C7H5IO | 232.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26260-02-6
Molecular Formula C7H5IO
Molecular Weight (g/mol) 232.02
MDL Number MFCD00039570
InChI Key WWKKTHALZAYYAI-UHFFFAOYSA-N
Synonym benzaldehyde, 2-iodo, o-iodobenzaldehyde, iodobenzaldehyde, benzaldehyde,iodo, 2-iodobenzenealdehyde, pubchem3914, acmc-209upi, 2-iodobenzaldehyde, ksc203m8l
PubChem CID 643439
IUPAC Name 2-iodobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)I

Specifications

Melting Point 36°C to 37°C
Boiling Point 102°C (3 mmHg)
Quantity 1 g
Beilstein 1927296
Sensitivity Air and light sensitive
Formula Weight 232.02
Percent Purity 98%
Chemical Name or Material 2-Iodobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.