missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzyl alcohol, 98+%

Product Code. 11328156
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11328156 25 g 25g
11318156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328156 Supplier Thermo Scientific Alfa Aesar Supplier No. L15126.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5159-41-1 | C7H7IO | 234.036 g/mol

2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1?-iodo-2?-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobenzyl alcohol was used in the synthesis of substituted seven-membered lactones, 2-[(E)-(1′-iodo-2′-propenyl)]benzyl alcohol2 and 2,3-diphenyl-1-indenone.

Solubility
Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5159-41-1
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00004610
InChI Key WZCXOBMFBKSSFA-UHFFFAOYSA-N
Synonym 2-iodobenzyl alcohol, 2-iodophenyl methanol, o-iodobenzyl alcohol, 2-iodobenzylalcohol, benzenemethanol, 2-iodo, 2-iodobenzylic alcohol, 2-jod-benzylalkohol, o-iodo benzylalcohol, 2-iodo-benzyl alcohol
PubChem CID 107629
IUPAC Name (2-iodophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)I

Specifications

Melting Point 89°C to 92°C
Quantity 25 g
Beilstein 2079487
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 234.04
Percent Purity 99%
Chemical Name or Material 2-Iodobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.