Learn More
2-Iodopropane (Stabilized with Copper), TRC
Product Code. 30004146
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004146 50 g 50g
30004030 1 g 1g
30004284 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30004146 Supplier Toronto Research Chemicals Supplier No. TRCI71879550G
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75-30-9
Molecular Formula C3 H7 I
Molecular Weight (g/mol) 169.99
Synonym Propane, 2-iodo- (8CI, 9CI, ACI), 2-Iodopropane (ACI), 1-Methylethyl iodide, 2-Propyl iodide, Isopropyl iodide, sec-Propyl iodide
IUPAC Name 2-iodopropane
SMILES CC(C)I
Specifications
Quantity 50 g
InChI Formula InChI=1S/C3H7I/c1-3(2)4/h3H,1-2H3
Formula Weight 169.9592
Chemical Name or Material 2-Iodopropane