missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-iso-Propylphenol-d12, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Isopropylphenol-D12 |
| CAS | 2708278-21-9 |
| Molecular Formula | C9 D12 O |
| Quantity | 0.25 g |
| Synonym | 2-iso-Propylphenol-d12, 2-[1-(Methyl-d3)ethyl-1,2,2,2-d4]phenol-O,3,4,5,6-d5, 2-(Propan-2-yl-d7)phenol-d5, 2-Isopropylphenol-D12, o-Hydroxycumene-d12, o-Isopropylphenol-d12 |
| InChI Formula | InChI=1 S/C9H12O/c1-7(2)8-5-3-4-6-9(8)10/h3-7,10 H,1-2H3/i1D3,2D3,3 D,4 D,5 D,6 D,7 D/hD |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c([2 H])c([2 H])c1C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-5-deuteriooxy-6-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)benzene |
| Molecular Weight (g/mol) | 148.265 |
| Formula Weight | 148.164 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction