missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylphenyl Diphenyl Phosphate, TRC

Product Code. 30134170
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134170 10 g 10g
30005202 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134170 Supplier Toronto Research Chemicals Supplier No. TRCI87287510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64532-94-1
Molecular Formula C21H21O4P
Molecular Weight (g/mol) 368.36
Synonym Phosphoric Acid 2-(1-Methylethyl)phenyl Diphenyl Ester, Diphenyl 2-Isopropylphenyl Phosphate, o-Isopropylphenyl Diphenyl Phosphate;
SMILES O=P(OC=1C=CC=CC1)(OC=2C=CC=CC2)OC=3C=CC=CC3C(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C21H21O4P/c1-17(2)20-15-9-10-16-21(20)25-26(22,23-18-11-5-3-6-12-18)24-19-13-7-4-8-14-19/h3-17H,1-2H3
Chemical Name or Material 2-Isopropylphenyl Diphenyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.