missing translation for 'onlineSavingsMsg'
Learn More

2-(Methacryloyloxy)ethyltrimethylammonium chloride, 72% aq. soln., stab. with 150-200 ppm 4-methoxyphenol

Product Code. 11383117
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383117 500 g 500g
11373117 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383117 Supplier Thermo Scientific Alfa Aesar Supplier No. L13523.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5039-78-1 | C9H18ClNO2 | 207.698 g/mol

2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methacryloyloxy)ethyltrimethylammonium chloride is a zirconium, aluminum and organic surface treated multipurpose rutile titanium dioxide pigment, it has narrow-size material distribution, good whiteness and high weather resistance. It is widely used in coating, plastics, paints, rubber and ink production, leather, cosmetics, soaps, plastics and in paper industry.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5039-78-1
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00060097
InChI Key RRHXZLALVWBDKH-UHFFFAOYSA-M
Synonym polyquaternium-37, 2-methacryloyloxy-n,n,n-trimethylethanaminium chloride, unii-pp88r88k3o, methacrylatoethyl trimethyl ammonium chloride, 2-methacryloyloxy ethyl trimethylammonium chloride, 2-methacryloyloxy ethyl trimethylammonium, methacryloxyethyltrimethyl ammonium chloride, ethanaminium, n,n,n-trimethyl-2-2-methyl-1-oxo-2-propenyl oxy-, chloride
PubChem CID 78738
IUPAC Name trimethyl-[2-(2-methylprop-2-enoyloxy)ethyl]azanium;chloride
SMILES CC(=C)C(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point -25°C
Density 1.105
Boiling Point >100°C
Refractive Index 1.466
Quantity 500 g
Beilstein 8168395
Solubility Information It is soluble in water.
Formula Weight 207.7
Concentration or Composition (by Analyte or Components) 72% aq. soln.
Chemical Name or Material 2-(Methacryloyloxy)ethyltrimethylammonium chloride, stabilized with 150 to 200ppm 4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.