missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-nitropyridine, 97%, Thermo Scientific™

Product Code. 11340817
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340817 50 g 50g
10476943 10 g 10g
10546334 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340817 Supplier Thermo Scientific Maybridge Supplier No. SPB06058EE
 Restricted Item
Estimated Shipment: 23-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 20265-35-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00023459
InChI Key WZNQCVOSOCGWJG-UHFFFAOYSA-N
Synonym 2-methoxy-3-nitro-pyridine, pyridine, 2-methoxy-3-nitro, 2-methoxy-3-nitropyridin, pubchem6593, maybridge1_008636, acmc-1cdn9, 2-methoxy-3-nitropyridine?, ksc494o8j, 2-methoxy-3-nitropyridine, 2-methoxy-3-nitropyridine 1g
PubChem CID 253219
IUPAC Name 2-methoxy-3-nitropyridine
SMILES COC1=C(C=CC=N1)[N+](=O)[O-]

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Methoxy-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.