Learn More
2-Methoxy-4-(prop-1-en-1-yl)phenol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 97-54-1 |
| Molecular Formula | C10H12O2 |
| MDL Number | MFCD00009285 |
| Quantity | 1 kg |
| SMILES | OC1=CC=C(/C=C/C)C=C1OC |
| Molecular Weight (g/mol) | 164.20 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Methoxy-4-(prop-1-en-1-yl)phenol |