missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-nitropyridine, 97%, Thermo Scientific™

Product Code. 15285958 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15285958 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15285958 Supplier Thermo Scientific Acros Supplier No. 125990050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5446-92-4
Melting Point 108.0°C to 111.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Molecular Formula C6H6N2O3
Linear Formula CH3O(C5H3N)NO2
MDL Number MFCD00006263
Quantity 5 g
Beilstein 21,II,33
Synonym 2-methoxy-5-nitro-pyridine, 5-nitro-2-methoxypyridine, 6-methoxy-3-nitropyridine, pyridine, 2-methoxy-5-nitro, methoxy 2--5-nitropyridine, 2-methoxy-5-nitropyridin, pubchem1207, acmc-1art3, 2-methoxy-5-nitro pyridine, 5-nitro-2-methoxy pyridine
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in chlorinated hydrocarbons,, insoluble in hexane
InChI Key WUPLOZFIOAEYMG-UHFFFAOYSA-N
SMILES COC1=CC=C(C=N1)[N+]([O-])=O
IUPAC Name 2-methoxy-5-nitropyridine
Molecular Weight (g/mol) 154.13
PubChem CID 79525
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-nitropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxy-5-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.