missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-d3-ethanol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyethanol-D3 (methoxy-D3) |
| CAS | 97840-77-2 |
| Molecular Formula | C3 D3 H5 O2 |
| Quantity | 0.5 g |
| Synonym | Ethanol, 2-(methoxy-d3)- (9 CI), 2-Methoxy-d3-ethanol, 2-(Methoxy-d3)ethanol, 2-Methoxyethanol-d3, 2-Methoxyethanol-D3 (methoxy-D3), Ethylene Glycol Monomethyl-d3 Ether |
| InChI Formula | InChI=1 S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])OCCO |
| IUPAC Name | 2-(trideuteriomethoxy)ethanol |
| Molecular Weight (g/mol) | 79.1129 |
| Formula Weight | 79.0713 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction