Learn More
2-Methoxybenzonitrile, 98%
Product Code. 11374534
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11374534 5 g 5g
11384534 25 g 25g
11394534 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11374534 Supplier Thermo Scientific Alfa Aesar Supplier No. L01293.06
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
CAS: 6609-56-9 | C8H7NO | 133.15 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6609-56-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001783
InChI Key FSTPMFASNVISBU-UHFFFAOYSA-N
Synonym 2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile
PubChem CID 81086
IUPAC Name 2-methoxybenzonitrile
SMILES COC1=CC=CC=C1C#N
Specifications
Density 1.093
Boiling Point 256°C
Flash Point >110°C (230°F)
Odor Almond-like
Refractive Index 1.5465
Quantity 5 g
UN Number UN3276
Beilstein 2207134
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 2-Methoxybenzonitrile