missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxyethoxymethyl Chloride, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxyethoxymethyl Chloride |
| CAS | 3970-21-6 |
| Molecular Formula | C4H9ClO2 |
| Quantity | 5 g |
| Synonym | 1-(Chloromethoxy)-2-methoxyethane, 2,5-Dioxahexyl Chloride, 2-(Chloromethoxy)ethoxymethane, 2-Methoxyethoxymethyl chloride, Chloromethyl 2-Methoxyethyl Ether, Methoxyethoxymethyl Chloride, beta-Methoxyethoxymethyl Chloride, MEM Chloride, MEM-Cl |
| InChI Formula | InChI=1S/C4H9ClO2/c1-6-2-3-7-4-5/h2-4H2,1H3 |
| SMILES | COCCOCCl |
| IUPAC Name | 1-(chloromethoxy)-2-methoxyethane |
| Molecular Weight (g/mol) | 124.57 |
| Formula Weight | 124.0291 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction