missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethoxymethyl Chloride, TRC

Product Code. 30472759
Click to view available options
Quantity:
10 g
Unit Size:
10g
This item is not returnable. View return policy

Product Code. 30472759

Brand: Toronto Research Chemicals TRCM26253010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxyethoxymethyl Chloride
CAS 3970-21-6
Molecular Formula C4H9ClO2
Quantity 10 g
Synonym 1-(Chloromethoxy)-2-methoxyethane, 2,5-Dioxahexyl Chloride, 2-(Chloromethoxy)ethoxymethane, 2-Methoxyethoxymethyl chloride, Chloromethyl 2-Methoxyethyl Ether, Methoxyethoxymethyl Chloride, beta-Methoxyethoxymethyl Chloride, MEM Chloride, MEM-Cl
InChI Formula InChI=1S/C4H9ClO2/c1-6-2-3-7-4-5/h2-4H2,1H3
SMILES COCCOCCl
IUPAC Name 1-(chloromethoxy)-2-methoxyethane
Molecular Weight (g/mol) 124.57
Formula Weight 124.0291
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.