missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)benzeneboronic acid, 97%

Product Code. 15459627
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459627 250 mg 250mg
15449627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459627 Supplier Thermo Scientific Alfa Aesar Supplier No. H52858.MD
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 126617-98-9 | C8H11BO3 | 165.983 g/mol

2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methoxymethyl)benzeneboronic acid used as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126617-98-9
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD03412110
InChI Key JHPPAFXQJZJGEP-UHFFFAOYSA-N
Synonym 2-methoxymethylphenylboronic acid, 2-methoxymethylbenzeneboronic acid, 2-methoxymethyl phenyl boronic acid, 2-methoxymethyl phenylboronic acid, 2-methoxymethyl benzeneboronic acid, 2-methoxymethylphebylboronic acid, boronic acid, 2-methoxymethyl phenyl, pubchem7997
PubChem CID 3759408
IUPAC Name [2-(methoxymethyl)phenyl]boronic acid
SMILES B(C1=CC=CC=C1COC)(O)O

Specifications

Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 165.99
Percent Purity 97%
Chemical Name or Material 2-(Methoxymethyl)benzeneboronic acid
compliance-icons
Product Identifier
  • 2-(Methoxymethyl)benzeneboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-(Methoxymethyl)phenylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.