missing translation for 'onlineSavingsMsg'
Learn More

2-(Methoxymethyl)oxirane, TRC

Product Code. 30476805
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476805 1 g 1g
30465158 5 g 5g
30469315 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30476805 Supplier Toronto Research Chemicals Supplier No. TRCM3299041G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-37-0
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
Synonym (Methoxymethyl)oxirane, (±)-Glycidyl Methyl Ether, 1,2-Epoxy-3-methoxypropane, 2,3-Epoxypropyl Methyl Ether, 3-Methoxy-1,2-epoxypropane, 3-Methoxypropylene Oxide, Denacol EX 131, Epiol M, Glycidol Methyl Ether, Glycidyl Methyl Ether, Methyl Glycidyl Ether, NSC 86950
IUPAC Name 2-(methoxymethyl)oxirane
SMILES COCC1CO1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H8O2/c1-5-2-4-3-6-4/h4H,2-3H2,1H3
Formula Weight 88.0524
Chemical Name or Material 2-(Methoxymethyl)oxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.