missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenylacetonitrile, 98%

Product Code. 11485847
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485847 50 g 50g
11495847 250 g 250g
11475847 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485847 Supplier Thermo Scientific Alfa Aesar Supplier No. B24400.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7035-03-2 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7035-03-2
MDL Number MFCD00001902

Specifications

Melting Point 66°C to 70°C
Boiling Point 141°C to 143°C (15 mmHg)
Flash Point 129°C (264°F)
Molecular Formula C9H9NO
Quantity 50 g
UN Number UN3439
Beilstein 2087748
Synonym 2-methoxyphenyl acetonitrile, 2-methoxyphenylacetonitrile, 2-methoxybenzyl cyanide, 2-2-methoxyphenyl acetonitrile, benzeneacetonitrile, 2-methoxy, o-methoxyphenylacetonitrile, o-methoxybenzyl cyanide, o-methoxyphenyl acetonitrile, 2-2-methoxyphenyl ethanenitrile, 2-methoxybenzylcyanid
InChI Key DWJKILXTMUGXOU-UHFFFAOYSA-N
SMILES COC1=CC=CC=C1CC#N
IUPAC Name 2-(2-methoxyphenyl)acetonitrile
Molecular Weight (g/mol) 147.18
PubChem CID 81496
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 2-Methoxyphenylacetonitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.